Anti-STC2 Chicken Polyclonal Antibody
Catalog # 89264-090
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Stanniocalcin 2
- Antigen Symbol:STC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- Isotype:IgY
- Reactivity:Human
- Western Blot:Yes
- Size:50 μg
- Gene ID:8614
- Antigen Synonyms:stanniocalcin-related protein|STCRP|stanniocalcin-2|STC-2|STC-related protein
- Storage Buffer:Phosphate-Buffered Saline containing no preservatives
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptide - human Stanniocalcin-2
- Purification:Affinity purified with antigen
- Cat. No.:89264-090
- Supplier no.:GTX82231
Specifications
About this item
Chicken polyclonal to Stanniocalcin-2
WB (only tested on fusion protein): Use at a dilution of 1:2000. Colorimetric method for signal development.
Type: Primary
Antigen: STC2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype:
Reactivity: Human