Anti-MDH1 Rabbit Polyclonal Antibody
Catalog # 89302-992
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Malate Dehydrogenase 1
- Antigen Symbol:MDH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Gel Shift:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:400 μl
- Gene ID:4190
- Antigen Synonyms:MOR2|MDH-s|MGC:1375|cytoplasmic|HEL-S-32|Malate dehydrogenase|EC=1.1.1.37|Cytosolic malate dehydrogenase|MDHA
- Storage Buffer:PBS with 0.09% (W/V) sodium azide
- Storage Temperature:Store at at 2-8°C for up to 6 months. For long term storage, aliquot and store at -20 °C. Avoid repeated freeze/thaw cycles.
- Concentration:0.25mg/ml
- Immunogen:KLH conjugated synthetic peptide between 293~322 amino acids from the C-terminal region of human MDH1.
- Purification:Prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis against PBS.
- Cat. No.:89302-992
- Supplier no.:GTX81955
Specifications
About this item
Rabbit polyclonal to MDH1 (C-term)
Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:50-100IHC: Use at 1:50-100FACS: Use at 1:10-50Optimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: MDH1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human