To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:CDP-Diacylglycerol-inositol 3-phosphatidyltransferase
- Antigen Symbol:CDIPT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Gel Shift:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 μl
- Environmentally Preferable:
- Gene ID:10423
- Antigen Synonyms:PIS1|PIS
- Storage Buffer:PBS with 0.09% (W/V) sodium azide
- Storage Temperature:Store at at 2-8°C for up to 6 months. For long term storage, aliquot and store at -20 °C. Avoid repeated freeze/thaw cycles.
- Concentration:0.25mg/ml
- Immunogen:KLH conjugated synthetic peptide selected from the Center region of human CDIPT.
- Purification:Purified through a protein A column, followed by peptide affinity purification.
- Cat. No.:89300-832
- Supplier no.:GTX80599
Specifications
About this item
Rabbit polyclonal to CDIPT (Internal)
Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500IHC: Use at 1:50-100FACS: Use at 1:10-50Optimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: CDIPT (CDP-diacylglycerol--inositol 3-phosphatidyltransferase)
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: