Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-RAN Rabbit Polyclonal Antibody
Anti-RAN Rabbit Polyclonal Antibody
Catalog # 89300-402
Supplier:  Genetex
Anti-RAN Rabbit Polyclonal Antibody
Catalog # 89300-402
Supplier:  Genetex
Supplier Number:  GTX80345

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    RAs-related Nuclear protein
  • Antigen Symbol:
    RAN
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Yeast
  • Western Blot:
    Yes
  • Size:
    50 μl
  • Gene ID:
    5901
  • Antigen Synonyms:
    OK/SW-cl.81|GTP-binding nuclear protein Ran|RAN|ARA24|GTPase Ran|Androgen receptor-associated protein 24|Ras-like protein TC4
  • Storage Buffer:
    Unpurified antiserum containing 0.1% sodium azide 
  • Storage Temperature:
    Store at 2-8°C. For extended storage, aliquot and store at –20 °C. Avoid repeated freeze-thaw cycles.
  • Immunogen:
    Purified recombinant GST-Gsp1p
  • Cat. No.:
    89300-402
  • Supplier no.:
    GTX80345

Specifications

About this item

Rabbit Polyclonal antibody to Ran (Gsp1p)

Recommended Starting Dilutions:Use at 1:1000Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: RAN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Yeast