Anti-POLR2A Mouse Monoclonal Antibody [clone: 4H8 ]
Catalog # 89280-876
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Polymerase (Rna) Ii (Dna Directed) Polypeptide A, 220Kda
- Antigen Symbol:POLR2A
- Clonality:Monoclonal
- Clone:4H8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:S. cerevisiae,Human
- Western Blot:Yes
- Size:50 μl
- Gene ID:5430
- Antigen Synonyms:hsRPB1|RPOL2|POLRA|RPBh1|RpIILS|RPO2|RPB1|POLR2|hRPB220
- Storage Buffer:PBS + 0.1% sodium azide
- Storage Temperature:Store at 2-8°C. For extended storage, aliquot and store at –20 °C. Avoid repeated freeze-thaw cycles.
- Immunogen:10 repeats of synthetic peptide: YSPTSPS using chemically synthesized phosphor-Ser5
- Purification:Protein A purified
- Cat. No.:89280-876
- Supplier no.:GTX80341
Specifications
About this item
Mouse monoclonal antibody [4H8] to RPB1
Optimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: POLR2A
Clonality: Monoclonal
Clone: 4H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, S. C. elegansrevisiae, Medaka