Anti-gp91phox Mouse Monoclonal Antibody [clone: 54.1 ]
Catalog # 89300-372
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Cytochrome B-245, Beta Polypeptide
- Antigen Symbol:gp91phox
- Clonality:Monoclonal
- Clone:54.1
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:50 μl
- Gene ID:1536
- Antigen Synonyms:GP91PHOX|p91-PHOX|AMCBX2|CGD|IMD34|NOX2|GP91-PHOX|GP91-1
- Storage Buffer:PBS + 0.1% sodium azide
- Storage Temperature:Store at 2-8°C. For extended storage, aliquot and store at –20 °C. Avoid repeated freeze-thaw cycles.
- Concentration:1mg/ml
- Immunogen:Partially Purified human neutrophil flavocytochrome b (heparin Ultrogel/lectin affinity purification in Triton X-100)
- Purification:Protein G
- Cat. No.:89300-372
- Supplier no.:GTX80327
Specifications
About this item
Mouse Monoclonal antibody [54.1] to gp91phox
Recommended Starting Dilutions:For WB: Use at 1:1000Optimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: CYBB
Clonality: Monoclonal
Clone: 54.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human