Anti-TNFRSF10B Mouse Monoclonal Antibody [clone: DR5-01-1]
Catalog # 89284-662
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Tumor Necrosis Factor Receptor Superfamily Member 10B
- Antigen Symbol:TNFRSF10B
- Clonality:Monoclonal
- Clone:DR5-01-1
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 μg
- Gene ID:8795
- Antigen Synonyms:Tumor necrosis factor receptor superfamily member 10B|TRAIL receptor 2|TRAILR2|TRAIL-R2|TRICK2|KILLER|Death receptor 5|ZTNFR9|TNFRSF10B|CD_antigen=CD262|TNF-related apoptosis-inducing ligand receptor 2|DR5
- Storage Buffer:The reagent is provided in phosphate buffered saline (PBS) containing 15 mM sodium azide and 0.2% (w/v) high-grade protease free Bovine Serum Albumin (BSA) as a stabilizing agent.
- Storage Temperature:Store in the dark at 2-8°C. Do not freeze. Do not use after expiration date stamped on vial label. Short-term exposure to room temperature should not affect the quality of the reagent.
- Immunogen:Recombinant fusion protein of human IgG heavy chain and extracellular domain of DR5.
- Cat. No.:89284-662
- Supplier no.:GTX80180
Specifications
About this item
Mouse monoclonal antibody [DR5-01-1] to DR5
Recommended Starting Dilutions:FACS: Use at 1-10 µg/mL. Positive Control: JURKAT human peripheral blood leukemia T cell lineOptimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: TNFRSF10B
Clonality: Monoclonal
Clone: DR5-01-1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human