Anti-TCRA +TCRB Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: IP26]
Catalog # 89298-946
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:T-cell Receptor Alpha And Beta
- Antigen Symbol:TCRA +TCRB
- Clonality:Monoclonal
- Clone:IP26
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Reactivity:Human
- Size:100 Tests
- Antigen Synonyms:Tcrb|FLJ22602|TCRA|MGC71411|MGC117436|T-cell receptor|MGC23964|TCRD|TCR beta|MGC22624|alpha polypeptide|TRA|Tcra|T-cell antigen receptor
- Cat. No.:89298-946
- Supplier no.:GTX80083
Specifications
About this item
Type: Primary
Antigen: TCRA +TCRB
Clonality: Monoclonal
Clone: IP26
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human