Anti-TCR Valpha2 Mouse Monoclonal Antibody [clone: F1]
Catalog # 89292-510
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:T-cell Receptor V alpha 2
- Antigen Symbol:TCR VA2
- Clonality:Monoclonal
- Clone:F1
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Primates,Human,Mouse
- Size:200 test
- Gene ID:6955
- Antigen Synonyms:TCRA|IMD7|TRA@|TCRD|TRAC
- Storage Buffer:PBS containing 0.1 % sodium azide and 0.5 % BSA
- Storage Temperature:4 °C
- Concentration:0.15 mg/ml
- Immunogen:Human TCR V?2
- Purification:Protein A
- Cat. No.:89292-510
- Supplier no.:GTX79225
Specifications
About this item
Mouse monoclonal antibody [F1] to TCR V alpha 2
Recommended Starting Dilutions: FACS: 1:10 - 1:100 IHC: 1:10 - 1:100 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: TCR Valpha2
Clonality: Monoclonal
Clone: F1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Primate