Anti-PRKAB1 Rabbit Polyclonal Antibody
Catalog # 89276-876
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Protein Kinase, AMP-Activated, beta 1 Non-Catalytic Subunit
- Antigen Symbol:PRKAB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:5564
- Antigen Synonyms:AMPK|HAMPKb
- Storage Buffer:PBS (without Mg2+and Ca2+), pH 7.4, 150mM NaCl,0.02% sodium azide and 50% glycerol.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Purification:Affinity purified with antigen
- Cat. No.:89276-876
- Supplier no.:GTX79141
Specifications
About this item
Rabbit polyclonal antibody to AMPK beta 1 (phospho Ser182)
Suggested starting dilutions are as follows. WB: 1:500-1:1000,IHC: 1:50-1:100,ELISA: 1:4000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: PRKAB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat