Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-JUNB Rabbit Polyclonal Antibody
  89276-652
 :  Genetex
Anti-JUNB Rabbit Polyclonal Antibody
  89276-652
 :  Genetex
 :  GTX79028

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Jun B Transcription Factor
  • Antigen Symbol:
    JUNB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 μg
  • Gene ID:
    3726
  • Antigen Synonyms:
    AP-1|activator protein 1|Jun B Transcription Factor|Transcription factor jun-BGene Symbols: JUNB|JUNB
  • Modification:
    phospho-Ser79
  • Storage Buffer:
    PBS (without Mg2+and Ca2+), pH 7.4, 150mM NaCl, 0.02%sodium azide and 50% glycerol.
  • Storage Temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Purification:
    Affinity purified with antigen
  • Cat. No.:
    89276-652
  • Supplier no.:
    GTX79028

 

 

Rabbit polyclonal antibody to JunB (phospho S79)

Suggested starting dilutions are as follows. WB: 1:500-1:1000,IHC: 1:50-1:100. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: JUNB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat