Anti-ICAM1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 1H4]
89287-148
: Genetex
- Antibody Type:Primary
- Antigen Name:Intercellular adhesion molecule 1
- Antigen Symbol:ICAM1
- Clonality:Monoclonal
- Clone:1H4
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Size:100 test
- Gene ID:3383
- Antigen Synonyms:CD54|VNRA|VTNR
- Storage Buffer:The reagent is provided in phosphate buffered saline (PBS) containing 15 mM sodium azide and 0.2% (w/v) high-grade protease free Bovine Serum Albumin (BSA) as a stabilizing agent.
- Storage Temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze. Keep protected from prolonged exposure to light.
- Immunogen:Raji cells and spleen cells fused with NS1 cells
- Cat. No.:89287-148
- Supplier no.:GTX78389
Monoclonal Antibody to CD54 (Human) Phycoerythrin (PE)
The reagent is designed for Flow Cytometry analysis of human blood cells using 20 ul reagent / 100 ul of whole blood or 106 cells in a suspension.The content of a vial (2 ml) is sufficient for 100 tests.
Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone: 1H4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human