Anti-CD10 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: LT10]
Catalog # 89287-142
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Neprilysin
- Antigen Symbol:CD10
- Clonality:Monoclonal
- Clone:LT10
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 test
- Gene ID:4311
- Antigen Synonyms:Calla|Common Acute Lymphoblastic Leukemia Antigen|Nep|Mme|Cd10|Membrane Metallo-Endopeptidase|Cluster Of Differentiation 10|Neutral Endopeptidase
- Storage Buffer:The reagent is provided in phosphate buffered saline (PBS) containing 15 mM sodium azide and 0.2% (w/v) high-grade protease free Bovine Serum Albumin (BSA) as a stabilizing agent.
- Storage Temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze. Keep protected from prolonged exposure to light.
- Immunogen:mouse NALM-6 leukemia pre-B cell line (tissue/cell preparation)
- Cat. No.:89287-142
- Supplier no.:GTX78377
Specifications
About this item
Monoclonal Antibody to CD10 (Human) Phycoerythrin (PE)
The reagent is designed for Flow Cytometry analysis of human blood cells using 20 ul reagent / 100 ul of whole blood or 106 cells in a suspension.The content of a vial (2 ml) is sufficient for 100 tests.
Type: Primary
Antigen: CD10
Clonality: Monoclonal
Clone: LT10
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human