Anti-NTAL Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: NAP-07]
Catalog # 89280-838
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Linker for activation of T cells Family, member 2
- Antigen Symbol:NTAL
- Clonality:Monoclonal
- Clone:NAP-07
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Size:100 μg
- Gene ID:7462
- Antigen Synonyms:LAB
- Storage Buffer:The reagent is provided in phosphate buffered saline (PBS) containing 15 mM sodium azide and 0.2% (w/v) high-grade protease free Bovine Serum Albumin (BSA) as a stabilizing agent.
- Storage Temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze. Keep protected from prolonged exposure to light.
- Concentration:0.1 mg/ml
- Immunogen:Recombinant cytoplasmic domain (aa 91-244) of human NTAL.
- Cat. No.:89280-838
- Supplier no.:GTX78366
Specifications
About this item
Monoclonal Antibody to NTAL/LAB Phycoerythrin (PE)
The reagent is designed for Flow Cytometry analysis.Suggested working dilution is 1:30. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.
Type: Primary
Antigen: NTAL
Clonality: Monoclonal
Clone: NAP-07
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse