Anti-CD46 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-258]
Catalog # 89287-138
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:CD46 molecule, complement regulatory protein
- Antigen Symbol:CD46
- Clonality:Monoclonal
- Clone:MEM-258
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 test
- Gene ID:4179
- Antigen Synonyms:MIC10|MCP|TRA2.10|AHUS2|TLX
- Storage Buffer:The reagent is provided in phosphate buffered saline (PBS) containing 15 mM sodium azide and 0.2% (w/v) high-grade protease free Bovine Serum Albumin (BSA) as a stabilizing agent.
- Storage Temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze. Keep protected from prolonged exposure to light.
- Immunogen:T cell line HPB-ALL.
- Cat. No.:89287-138
- Supplier no.:GTX78353
Specifications
About this item
Monoclonal Antibody to CD46 (Human) Phycoerythrin (PE)
The reagent is designed for Flow Cytometry analysis of human blood cells using 20 ul reagent / 100 ul of whole blood or 106 cells in a suspension.The content of a vial (2 ml) is sufficient for 100 tests.
Type: Primary
Antigen: CD46
Clonality: Monoclonal
Clone: MEM-258
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: