Anti-PTPRC Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-28]
Catalog # 89287-126
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Protein Tyrosine Phosphatase Receptor Type C
- Antigen Symbol:PTPRC
- Clonality:Monoclonal
- Clone:MEM-28
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 test
- Gene ID:5788
- Antigen Synonyms:CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
- Storage Buffer:The reagent is provided in phosphate buffered saline (PBS) containing 15 mM sodium azide and 0.2% (w/v) high-grade protease free Bovine Serum Albumin (BSA) as a stabilizing agent.
- Storage Temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze. Keep protected from prolonged exposure to light.
- Immunogen:Human thymocytes and T lymphocytes.
- Cat. No.:89287-126
- Supplier no.:GTX78295
Specifications
About this item
Monoclonal Antibody to CD45 (Human) Phycoerythrin (PE) conjugated
The reagent is designed for Flow Cytometry analysis of human blood cells using 20 ul reagent / 100 ul of whole blood or 106 cells in a suspension.The content of a vial (2 ml) is sufficient for 100 tests.
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: MEM-28
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human