Anti-CD8 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-31]
89287-118
: Genetex
- Antibody Type:Primary
- Antigen Name:Cluster of differentiation 8
- Antigen Symbol:CD8
- Clonality:Monoclonal
- Clone:MEM-31
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Size:100 test
- Gene ID:925
- Antigen Synonyms:CD8alpha|p32|Leu-2|CD8a|MAL|CD8|CD8b|CD8beta
- Storage Buffer:The reagent is provided in phosphate buffered saline (PBS) containing 15 mM sodium azide and 0.2% (w/v) high-grade protease free Bovine Serum Albumin (BSA) as a stabilizing agent.
- Storage Temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze. Keep protected from prolonged exposure to light.
- Immunogen:Crude thymus membrane fraction.
- Cat. No.:89287-118
- Supplier no.:GTX78259
Monoclonal Antibody to CD8 (Human) Phycoerythrin (PE) conjugated
The reagent is designed for Flow Cytometry analysis of human blood cells using 20 _l reagent / 100 _l of whole blood or 106 cells in a suspension. The content of a vial (2 ml) is sufficient for 100 tests.
Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: MEM-31
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: