Anti-YEATS4 Rabbit Polyclonal Antibody
89263-644
: Genetex
- Antibody Type:Primary
- Antigen Name:YEATS Domain Containing 4
- Antigen Symbol:YEATS4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 μg
- Gene ID:8089
- Antigen Synonyms:GAS41|NUBI-1|4930573H17Rik|B230215M10Rik|YAF9
- Storage Buffer:PBS with 2% sucrose
- Storage Temperature:Store at -20 °C or below. Avoid repeat freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Polyclonal antibody produced in rabbits immunized with a synthetic peptide corresponding to a region of human YEATS4
- Purification:Rabbit IgG is purified by peptide affinity chromatography method.
- Cat. No.:89263-644
- Supplier no.:GTX77840
Rabbit Polyclonal Antibody to Glioma-Amplified Sequence-41 (YEATS4)
WB: Suggested dilution at 0.5 ug/ml in 5% skim milk / PBS buffer. Optimal dilutions should be determined by end-user.
Type: Primary
Antigen: YEATS4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human