Anti-HMGB2 Rabbit Polyclonal Antibody
89280-002
: Genetex
- Antibody Type:Primary
- Antigen Name:High-mobility Group Box 2
- Antigen Symbol:HMGB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 μg
- Gene ID:3148
- Antigen Synonyms:High mobility group protein B2|HMG-2 |synonyms: HMG2|High mobility group protein 2|name: HMGB2
- Storage Buffer:Phosphate-bufferd saline containing 2% sucrose
- Storage Temperature:Upon Receipt - Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Polyclonal antibody produced in rabbits immunized with a synthetic peptide corresponding to a region of human HMGB2
- Cat. No.:89280-002
- Supplier no.:GTX77816
Rabbit Polyclonal Antibody to HMGB2
For IHC-P: Use at a concentration of 4.0-8.0 ?g/ml. For WB: Use at a concentration of 2.0 ?g/ml in 5% skim milk/PBS buffer. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: HMGB2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human