Anti-TFCP2L1 Rabbit Polyclonal Antibody
89263-640
: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Transcription Factor CP2-like 1
- Antigen Symbol:TFCP2L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 μg
- Gene ID:29842
- Antigen Synonyms:CRTR-1|CRTR1|Transcription factor CP2-like protein 1|CP2-related transcriptional repressor 1|TFCP2L1|LBP9|LBP-9
- Storage Buffer:PBS with 2% sucrose
- Storage Temperature:Store at -20 °C or below. Avoid repeat freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Polyclonal antibody produced in rabbits immunized with a synthetic peptide corresponding to a region of human TFCP2L1
- Purification:Rabbit Ig G is purified by peptide affinity chromatography method.
- Cat. No.:89263-640
- Supplier no.:GTX77815
Rabbit Polyclonal Antibody to TFCP2L1
- WB: Suggested dilution at 1.0-5.0 ug/ml in 5% skim milk / PBS buffer. Optimal dilutions should be determined by end-user.
Type: Primary
Antigen: TFCP2L1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human