Anti-P53 Mouse Monoclonal Antibody [clone: DO-7]
Catalog # 89263-418
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Tumor protein p53
- Antigen Symbol:P53
- Clonality:Monoclonal
- Clone:DO-7
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western Blot:Yes
- Size:50 μg
- Gene ID:7157
- Antigen Synonyms:or transformation-related protein 53 (TRP53)|tumor suppressor p53|antigen NY-CO-13|P53|BCC7|TRP53|phosphoprotein p53|LFS1|tumor protein p53
- Storage Buffer:Phosphate buffered saline containing 0.09% Sodium Azide
- Storage Temperature:Store at +4 °C or at –20 °C if preferred. This product should be stored undiluted. Should this product contain a precipitate we recommend microcentrifugation before use.
- Immunogen:Recombinant human p53.
- Purification:Purified IgG prepared by affinity chromatography on Protein G
- Cat. No.:89263-418
- Supplier no.:GTX75255
Specifications
About this item
Mouse monoclonal antibody [DO-7] to p53
Recommended Starting Dilutions: Western Blotting: Max Dilution 1/1000 Flow Cytometry: 1/50 - 1/100 Immunohistology - Paraffin: Max Dilution 1/1000 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: P53
Clonality: Monoclonal
Clone: DO-7
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human