Anti-CCL9 Rabbit Polyclonal Antibody
Catalog # 89281-172
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Chemokine (C-C motif) Ligand 9
- Antigen Symbol:CCL9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:20308
- Antigen Synonyms:small inducible cytokine A10|macrophage inflammatory protein 1-gamma|small inducible cytokine A9|MIP-1 gamma|C-C motif chemokine 9|macrophage inflammatory protein-related protein 2
- Storage Buffer:Phosphate buffered saline. For long term storage, the addition of 0. 09% sodium azide is recommended. For functional studies, do not use azide.
- Storage Temperature:Prior to reconstitution store at +4 °C. Following reconstitution store at –20 °C. Store undiluted. Storage in frost-free freezers is not recommended. Should this product contain a precipitate we recommend microcentrifugation before use.
- Immunogen:Recombinant mouse MIP1 gamma
- Purification:Purified IgG was prepared from whole serum by affinity chromatography.
- Cat. No.:89281-172
- Supplier no.:GTX74083
Specifications
About this item
Rabbit polyclonal antibody to MIP1 gamma
Recommended Starting Dilutions: ELISA: Max Dilution 0.5 ug/ml Western Blotting: 0.1 ug/ml - 0.2 ug/ml Not tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: CCL9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: