To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:MCP5
- Antigen Symbol:MCP5 (Chemokine (C-C motif) ligand 12)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 μg
- Neutralisation:Yes
- Gene ID:20293
- Storage Buffer:Phosphate buffered saline. For long term storage, the addition of 0. 09% sodium azide is recommended. For functional studies, do not use azide.
- Storage Temperature:Prior to reconstitution store at +4 °C. Following reconstitution store at –20 °C. Store undiluted. Storage in frost-free freezers is not recommended. Should this product contain a precipitate we recommend microcentrifugation before use.
- Immunogen:Recombinant mouse MCP5
- Purification:Purified IgG was prepared from whole serum by affinity chromatography.
- Cat. No.:89281-170
- Supplier no.:GTX74079
Specifications
About this item
Rabbit polyclonal antibody to MCP5
Recommended Starting Dilutions: ELISA: Max Dilution 0.5 ug/ml Functional Assays: 2.0 ug/ml - 3.0 ug/ml Western Blotting: 0.1 ug/ml - 0.2 ug/ml Not tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: Ccl12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse