Anti-GPCR RTA Rabbit Polyclonal Antibody
Catalog # 89278-196
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Symbol:RTA
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Size:25 µg
- Cat. No.:89278-196
- Supplier no.:GTX71787
Specifications
About this item
Type: Primary
Antigen: GPCR RTA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human