Anti-GPCR MRGE (MAS-related GPR member E) Rabbit Polyclonal Antibody
Catalog # 89278-114
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Mas-Related Gpr, Member E
- Antigen Symbol:MRGE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:25 μg
- Gene ID:116534
- Antigen Synonyms:MGRF|MRGE|GPR167
- Storage Buffer:Phosphate buffered saline, PH 7.7 containing 0.01% sodium azide
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide - KLH conjugated
- Purification:Immunogen affinity purified.
- Cat. No.:89278-114
- Supplier no.:GTX71501
Specifications
About this item
Rabbit Polyclonal to Human G Protein - Coupled Receptor MRGE
IHC-P: 1/100 - 1/200. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: MRGPRE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human