Anti-TFE3 Rabbit Polyclonal Antibody
89306-618
: Genetex
- Antibody Type:Primary
- Antigen Name:Transcription Factor Binding to IGHM Enhancer 3
- Antigen Symbol:TFE3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:7030
- Antigen Synonyms:bHLHe33|TFEA|RCCP2|RCCX1
- Storage Buffer:saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Peptide sequence around aa.303~307 (L-D-V-Y-S) derived from Human TFE3.
- Purification:Antibodies were purified by affinity-chromatography using epitope-specific peptide.
- Cat. No.:89306-618
- Supplier no.:GTX50738
Rabbit Polyclonal antibody to TFE3
Suggested starting dilutions are as follows. WB: 1:500~1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: TFE3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse