Anti-CCNB1 Rabbit Polyclonal Antibody
Catalog # 89304-556
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Cyclin B1
- Antigen Symbol:CCNB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:891
- Antigen Synonyms:CCNB
- Modification:phospho-Ser147
- Storage Buffer:saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Peptide sequence around phosphorylation site of Serine 147 (A-F-S(p)-D-V) derived from Human Cyclin B1.
- Purification:Antibodies were purified by affinity-chromatography using epitope-specific phosphopeptide. Non-phospho specific antibodies were removed by chromatogramphy using non-phosphopeptide.
- Cat. No.:89304-556
- Supplier no.:GTX50351
Specifications
About this item
Rabbit Polyclonal antibody to Cyclin B1 (Phospho Ser147)
Suggested starting dilutions are as follows. WB: 1:500~1:1000, IHC-P: 1:50~1:100, ICC/IF: 1:100~1:200. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: CCNB1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human