Anti-HSP27 Rabbit Polyclonal Antibody
89304-410
: Genetex
- Antibody Type:Primary
- Antigen Name:Heat Shock Protein 27
- Antigen Symbol:HSP27
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:3315
- Antigen Synonyms:SRP27|HSP27|HSP28|HspB1|Hsp25|Hs.76067|HSPB1|HS.76067|DKFZp586P1322|HMN2B|CMT2F
- Modification:phospho-Ser82
- Storage Buffer:saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Peptide sequence around phosphorylation site of serine 82 (Q-L-S(p)-S-G) derived from Human HSP27.
- Purification:Antibodies were purified by affinity-chromatography using epitope-specific phosphopeptide. Non-phospho specific antibodies were removed by chromatogramphy using non-phosphopeptide.
- Cat. No.:89304-410
- Supplier no.:GTX50277
Rabbit Polyclonal antibody to HSP27 (Phospho Ser82)
Suggested starting dilutions are as follows. WB: 1:500~1:1000, IHC-P: 1:50~1:100, ICC/IF: . Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: HSP27
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human