To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Post-GPI attachment to Proteins 3
- Antigen Symbol:PGAP3
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 μg
- Gene ID:93210
- Antigen Synonyms:AGLA546|PERLD1|CAB2|PP1498|hCOS16
- Storage Buffer:Phosphate, pH7.2-7.4
- Storage Temperature:Product should be aliquoted and stored at -20∞C. Avoid repeated freeze/thaw cycles.
- Immunogen:Synthetic peptide
- Purification:Affinity purified using antigen.
- Cat. No.:89275-894
- Supplier no.:GTX47867
Specifications
About this item
FITC-conjugated Rabbit polyclonal antibody to Post-GPI attachment to proteins 3
Optimal working dilutions should be determined experimentally be the end user.
Type: Primary
Antigen: PGAP3
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human