Anti-APOBPEC3F Rabbit Polyclonal Antibody
Catalog # 89270-350
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Apolipoprotein B mRNA Editing Enzyme Catalytic Polypeptide-Like 3F
- Antigen Symbol:APOBPEC3F
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 μg
- Gene ID:200316
- Antigen Synonyms:KA6|ARP8|A3F|BK150C2.4.MRNA
- Storage Buffer:Phosphate buffered saline (PBS)
- Storage Temperature:Store at 4 °C for short-term use. For longer periods of storage, aliquot and store at –20 °C. Avoid repeat freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide
- Purification:Peptide Affinity Purified
- Cat. No.:89270-350
- Supplier no.:GTX47211
Specifications
About this item
Rabbit polyclonal antibody to APOBEC3F (C-terminal)
Recommended Dilutions: For Western Blot: Use at 1 ug/ml.
Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: APOBPEC3F
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human