Anti-CSPG5 (Chondroitin sulfate proteoglycan 5 (neuroglycan C)) Rabbit Polyclonal Antibody
Catalog # 89269-092
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Chondroitin sulfate proteoglycan 5
- Antigen Symbol:CSPG5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 μg
- Gene ID:10675
- Antigen Synonyms:NGC
- Storage Buffer:Phosphate buffered saline (PBS)
- Storage Temperature:Store at 4 °C for short-term use. For longer periods of storage, aliquot and store at –20 °C. Avoid repeat freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide
- Purification:Peptide Affinity Purified
- Cat. No.:89269-092
- Supplier no.:GTX46412
Specifications
About this item
Rabbit polyclonal antibody to CSPG5 (N-terminal)
Recommended Dilutions: For Western Blot: Use at 1 ug/ml.
Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: CSPG5 (Chondroitin sulfate proteoglycan 5 (neuroglycan C))
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: