Anti-RPRD1B Rabbit Polyclonal Antibody
Catalog # 89266-290
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Regulation of Nuclear Pre-mRNA Domain Containing 1B
- Antigen Symbol:RPRD1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 μg
- Gene ID:58490
- Antigen Synonyms:C20orf77|NET60|dJ1057B20.2|CREPT
- Storage Buffer:Phosphate buffered saline (PBS)
- Storage Temperature:Store at 4 °C for short-term use. For longer periods of storage, aliquot and store at –20 °C. Avoid repeat freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide
- Purification:Peptide Affinity Purified
- Cat. No.:89266-290
- Supplier no.:GTX44898
Specifications
About this item
Rabbit polyclonal antibody to RPRD1B (Middle)
Optimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: RPRD1B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human