Anti-CDXB Mouse Monoclonal Antibody [clone: B427M]
Catalog # 89280-748
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Symbol:Clostridium difficile toxin B
- Clonality:Monoclonal
- Clone:B427M
- ELISA:Yes
- Host:Mouse
- Reactivity:Bacteria
- Size:100 µg
- Cat. No.:89280-748
- Supplier no.:GTX44092
Specifications
About this item
Type: Primary
Antigen: CDXB
Clonality: Monoclonal
Clone: B427M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: