Anti-F3 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: CLB/TF-5]
Catalog # 89279-614
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Coagulation Factor 3
- Antigen Symbol:F3
- Clonality:Monoclonal
- Clone:CLB/TF-5
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 μg
- Gene ID:2152
- Antigen Synonyms:F3|TFA|Coagulation factor 3|CD142|TF
- Storage Buffer:TRIS buffered saline containing 0.09% Sodium Azide (NaN3) 1% Bovine Serum Albumin
- Storage Temperature:Store at +4 °C or at –20 °C if preferred. Storage in frost-free freezers is not recommended. Store undiluted. Photosensitive. Should this product contain a precipitate we recommend microcentrifugation before use.
- Purification:Purified IgG prepared by affinity chromatography on Protein A from tissue culture supernatant
- Cat. No.:89279-614
- Supplier no.:GTX43752
Specifications
About this item
Mouse monoclonal antibody [CLB/TF-5] to Tissue Factor
Type: Primary
Antigen: F3
Clonality: Monoclonal
Clone: CLB/TF-5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human