Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Cytomegalovirus Glycoprotein B Mouse Monoclonal Antibody [clone: 1-M-12]
Anti-Cytomegalovirus Glycoprotein B Mouse Monoclonal Antibody [clone: 1-M-12]
Catalog # 89280-736
Supplier:  Genetex
Anti-Cytomegalovirus Glycoprotein B Mouse Monoclonal Antibody [clone: 1-M-12]
Catalog # 89280-736
Supplier:  Genetex
Supplier Number:  GTX43527

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Cytomegalovirus Glycoprotein B
  • Antigen Symbol:
    CMV Glycoprotein B
  • Clonality:
    Monoclonal
  • Clone:
    1-M-12
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Virus
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Cat. No.:
    89280-736
  • Supplier no.:
    GTX43527

Specifications

About this item

Type: Primary
Antigen: Cytomegalovirus Glycoprotein B
Clonality: Monoclonal
Clone: 1-M-12
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus