Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CD68 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: ED1]
Anti-CD68 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: ED1]
Catalog # 89262-968
Supplier:  Genetex
Anti-CD68 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: ED1]
Catalog # 89262-968
Supplier:  Genetex
Supplier Number:  GTX43522
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    CD68 molecule
  • Antigen Symbol:
    CD68
  • Clonality:
    Monoclonal
  • Clone:
    ED1
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Rat
  • Size:
    100 μg
  • Gene ID:
    287435
  • Antigen Synonyms:
    macrosialin|CD68 antigen|GP110|scavenger receptor class D member 1|SCARD1|LAMP4|macrophage antigen CD68
  • Storage Buffer:
    Phosphate buffered saline containing 0.09% Sodium Azide 1% Bovine Serum Albumin
  • Storage Temperature:
    Store at +4 °C or at –20 °C if preferred. Store undiluted. Storage in frost-free freezers is not recommended. Photosensitive. Should this product contain a precipitate we recommend microcentrifugation before use.
  • Purification:
    Purified IgG prepared by affinity chromatography on Protein A from tissue culture supernatant
  • Cat. No.:
    89262-968
  • Supplier no.:
    GTX43522

Specifications

About this item

Mouse monoclonal antibody [ED1] to CD68

Type: Primary
Antigen: CD68
Clonality: Monoclonal
Clone: ED1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: