Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CD8 beta Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 341]
Anti-CD8 beta Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 341]
Catalog # 89285-708
Supplier:  Genetex
Anti-CD8 beta Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 341]
Catalog # 89285-708
Supplier:  Genetex
Supplier Number:  GTX43496

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    CD8 Beta
  • Antigen Symbol:
    CD8 beta
  • Clonality:
    Monoclonal
  • Clone:
    341
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Rat
  • Size:
    100 μg
  • Gene ID:
    24931
  • Storage Buffer:
    Phosphate buffered saline containing 0.09% Sodium Azide 1% Bovine Serum Albumin
  • Storage Temperature:
    Store at +4 °C for one month or at –20 °C for longer. This product should be stored undiluted. This product is photosensitive and should be protected from light. Should this product contain a precipitate we recommend microcentrifugation before use.
  • Purification:
    Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
  • Cat. No.:
    89285-708
  • Supplier no.:
    GTX43496

Specifications

About this item

Mouse monoclonal antibody [341] to CD8 beta

Recommended Starting Dilutions: Flow Cytometry: 1/2 - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD8 beta
Clonality: Monoclonal
Clone: 341
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: