Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CD117 (kit oncogene) Rat Monoclonal Antibody [clone: 2B8]
Anti-CD117 (kit oncogene) Rat Monoclonal Antibody [clone: 2B8]
Catalog # 89306-976
Supplier:  Genetex
Anti-CD117 (kit oncogene) Rat Monoclonal Antibody [clone: 2B8]
Catalog # 89306-976
Supplier:  Genetex
Supplier Number:  GTX43210

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    KIT Proto-oncogene Receptor Tyrosine Kinase
  • Antigen Symbol:
    KIT
  • Clonality:
    Monoclonal
  • Clone:
    2B8
  • Conjugation:
    PE (Phycoerythrin)
  • Flow Cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2b
  • Reactivity:
    Mouse
  • Size:
    50 test
  • Gene ID:
    16590
  • Antigen Synonyms:
    SCFR|CD117|PBT|C-Kit
  • Storage Buffer:
    Phosphate buffered saline containing 0.09% Sodium Azide 1% Bovine Serum Albumin 5% Sucrose
  • Storage Temperature:
    Store at +4°C. DO NOT FREEZE. This product should be stored undiluted. This product is photosensitive and should be protected from light. Should this product contain a precipitate we recommend microcentrifugation before use.
  • Purification:
    Purified IgG prepared by affinity chromatography on Protein A.
  • Cat. No.:
    89306-976
  • Supplier no.:
    GTX43210

Specifications

About this item

Rat monoclonal antibody [2B8] to c-Kit

Type: Primary
Antigen: CD117 (kit oncogene)
Clonality: Monoclonal
Clone: 2B8
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse