Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-CD200R (CD200R 1) Mouse Monoclonal Antibody [clone: OX108]
  89315-782
 :  Genetex
Anti-CD200R (CD200R 1) Mouse Monoclonal Antibody [clone: OX108]
  89315-782
 :  Genetex
 :  GTX43159

 

  • Antibody Type:
    Primary
  • Antigen Name:
    CD200 Receptor 1
  • Antigen Symbol:
    CD200R1
  • Clonality:
    Monoclonal
  • Clone:
    OX108
  • Conjugation:
    PE (Phycoerythrin)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    25 test
  • Gene ID:
    131450
  • Antigen Synonyms:
    MOX2R|HCRTR2|CD200R|OX2R
  • Storage Buffer:
    Phosphate buffered saline containing 0.09% Sodium Azide 1% Bovine Serum Albumin 5% Sucrose
  • Storage Temperature:
    Store at +4°C. DO NOT FREEZE. This product should be stored undiluted. This product is photosensitive and should be protected from light. Should this product contain a precipitate we recommend microcentrifugation before use.
  • Purification:
    Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
  • Cat. No.:
    89315-782
  • Supplier no.:
    GTX43159

 

 

Mouse monoclonal antibody [OX108] to CD200R

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD200R (CD200R 1)
Clonality: Monoclonal
Clone: OX108
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human