Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-O157 Mouse Monoclonal Antibody [clone: B835M]
Anti-O157 Mouse Monoclonal Antibody [clone: B835M]
  89280-732
 :  Genetex
Anti-O157 Mouse Monoclonal Antibody [clone: B835M]
  89280-732
 :  Genetex
 :  GTX43112
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    E. coli O157
  • Antigen Symbol:
    O157
  • Clonality:
    Monoclonal
  • Clone:
    B835M
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Bacteria
  • Size:
    100 µg
  • Cat. No.:
    89280-732
  • Supplier no.:
    GTX43112

 

 

Type: Primary
Antigen: O157
Clonality: Monoclonal
Clone: B835M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria