Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-CD314 Mouse Monoclonal Antibody [clone: 1D11]
  89274-802
 :  Genetex
Anti-CD314 Mouse Monoclonal Antibody [clone: 1D11]
  89274-802
 :  Genetex
 :  GTX43108

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Killer Cell Lectin-Like Receptor Subfamily K, Member 1
  • Antigen Symbol:
    CD314
  • Clonality:
    Monoclonal
  • Clone:
    1D11
  • Conjugation:
    PE (Phycoerythrin)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    100 test
  • Gene ID:
    22914
  • Antigen Synonyms:
    NKG2D
  • Storage Buffer:
    Phosphate buffered saline containing 0.09% Sodium Azide 1% Bovine Serum Albumin 5% Sucrose
  • Storage Temperature:
    Store at 4°C. DO NOT FREEZE. This product should be stored undiluted. This product is photosensitive and should be protected from light. Should this product contain a precipitate we recommend microcentrifugation before use.
  • Purification:
    Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant.
  • Cat. No.:
    89274-802
  • Supplier no.:
    GTX43108

 

 

Mouse monoclonal antibody [1D11] to NKG2D

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/5 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD314
Clonality: Monoclonal
Clone: 1D11
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: