Anti-CD34 Class II Mouse Monoclonal Antibody [clone: QBEND/10]
Catalog # 89274-800
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Cd34 Molecule
- Antigen Symbol:CD34 Class II
- Clonality:Monoclonal
- Clone:QBEND/10
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Cynomolgus monkey,Macaque,Human
- Size:100 test
- Gene ID:947
- Storage Buffer:Phosphate buffered saline containing 0.09% Sodium Azide 1% Bovine Serum Albumin
- Storage Temperature:Store at 4°C. DO NOT FREEZE. This product should be stored undiluted. This product is photosensitive and should be protected from light. Should this product contain a precipitate we recommend microcentrifugation before use.
- Purification:Purified IgG prepared by affinity chromatography on Protein A from tissue culture supernatant
- Cat. No.:89274-800
- Supplier no.:GTX43101
Specifications
About this item
Mouse monoclonal antibody [QBEND/10] to CD34 Class II
Type: Primary
Antigen: CD34 Class II
Clonality: Monoclonal
Clone: QBEND/10
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: