Anti-E. coli Shigatoxin (STX) 1 & 2 Mouse Monoclonal Antibody [clone: Mixed]
Catalog # 89316-810
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:E. coli Shigatoxin (STX) 1 + 2
- Antigen Symbol:E. coli Shigatoxin (STX) 1 + 2
- Clonality:Monoclonal
- Clone:Mixed
- ELISA:Yes
- Host:Mouse
- Reactivity:Bacteria
- Western Blot:Yes
- Size:200 µg
- Neutralisation:Yes
- Cat. No.:89316-810
- Supplier no.:GTX43048
Specifications
About this item
Type: Primary
Antigen: E. coli Shigatoxin (STX) 1 & 2
Clonality: Monoclonal
Clone: Mixed
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria