Anti-MRC1 Mouse Monoclonal Antibody [clone: 7-450]
Catalog # 89275-624
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Mannose Receptor, C type 1
- Antigen Symbol:MRC1
- Clonality:Monoclonal
- Clone:7-450
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Size:200 μg
- Gene ID:4360
- Antigen Synonyms:CD206|MMR
- Storage Buffer:Phosphate buffered saline containing 0.09% Sodium Azide (NaN3)
- Storage Temperature:Store at +4 °C or at –20 °C if preferred. Storage in frost-free freezers is not recommended. This product should be stored undiluted. Should this product contain a precipitate we recommend microcentrifugation before use.
- Purification:Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
- Cat. No.:89275-624
- Supplier no.:GTX42267
Specifications
About this item
Mouse monoclonal antibody [7-450] to Mannose Receptor
Recommended Starting Dilutions: Immunohistology - Frozen: 1/50 - 1/500 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: MRC1
Clonality: Monoclonal
Clone: 7-450
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human