Anti-Shank2 Mouse Monoclonal Antibody [clone: S23b-49]
89265-964
: Genetex
- Antibody Type:Primary
- Antigen Name:SH3 And Multiple Ankyrin Repeat Domains 2
- Antigen Symbol:SHANK2
- Clonality:Monoclonal
- Clone:S23b-49
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:171093
- Antigen Synonyms:AUTS17|CORTBP1|ProSAP1|SHANK|CTTNBP1|SPANK-3
- Storage Buffer:PBS pH7.4, 50% glycerol
- Storage Temperature:Aliquot and store at -20 C. Avoid repeated freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Fusion protein amino acids 84-309 of rat Shank2; accession number Q9QX74
- Purification:Protein G Purified
- Cat. No.:89265-964
- Supplier no.:GTX42006
Mouse monoclonal antibody [S23b-49] to PAN-SHANK
We recommend a working dilution of 1-10ug/mL (WB), 0.1-1.0ug/mL (Perox) (IHC/ICC), 1.0-10ug/mL (IF).
Type: Primary
Antigen: Shank2
Clonality: Monoclonal
Clone: S23b-49
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat