Anti-CD8 Alpha Mouse Monoclonal Antibody [clone: Du CD8-1]
Catalog # 89275-554
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD8 Alpha
- Clonality:Monoclonal
- Clone:Du CD8-1
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Reactivity:Duck
- Size:250 µg
- Cat. No.:89275-554
- Supplier no.:GTX41834
Specifications
About this item
Type: Primary
Antigen: CD8 Alpha
Clonality: Monoclonal
Clone: Du CD8-1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: