Anti-Er-Tr7 Rat Monoclonal Antibody [clone: ER-TR7]
Catalog # 89280-562
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Symbol:Er-Tr7
- Clonality:Monoclonal
- Clone:ER-TR7
- Host:Rat
- Reactivity:Mouse
- Size:250 µg
- Cat. No.:89280-562
- Supplier no.:GTX41261
Specifications
About this item
Type: Primary
Antigen: Er-Tr7
Clonality: Monoclonal
Clone: ER-TR7
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse