Anti-Nitrotyrosine Mouse Monoclonal Antibody [clone: 6B2-3G2]
Catalog # 89275-314
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Nitrotyrosine
- Antigen Symbol:Nitro-Tyr
- Clonality:Monoclonal
- Clone:6B2-3G2
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Western Blot:Yes
- Size:100 µg
- Cat. No.:89275-314
- Supplier no.:GTX39562
Specifications
About this item
Type: Primary
Antigen: Nitrotyrosine
Clonality: Monoclonal
Clone: 6B2-3G2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: [delete]