Anti-TNFA Rabbit Polyclonal Antibody
Catalog # 89276-074
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Tumor Necrosis Factor Alpha
- Antigen Symbol:TNFA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Bovine
- Western Blot:Yes
- Size:500 μg
- Gene ID:280943
- Antigen Synonyms:tumor necrosis factor ligand superfamily member 2|tumor necrosis factor alpha|DIF|monocyte-derived|Tnfa|cachectin|Tnfsf1a|TNFalpha|member 2|TNF-alpha|TNF-a|TNFSF2|macrophage-derived|TNF superfamily|TNF|tumor necrosis factor-alpha|APC1 protein
- Storage Buffer:Phosphate buffered saline
- Immunogen:Recombinant bovine TNF alpha.
- Purification:Purified IgG was prepared by affinity chromatography on protein G.
- Cat. No.:89276-074
- Supplier no.:GTX38917
Specifications
About this item
Rabbit polyclonal antibody to TNF alpha
Recommended Starting Dilutions:ELISA: 1 ug/ml - 10 ug/mlWestern Blotting: 1 ug/ml - 10 ug/mlNot tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: TNFA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine