To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Forkhead box protein J1
- Antigen Symbol:FOXJ1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:2302
- Antigen Synonyms:FOXJ1|HFH-4|HFH4|FKHL13|Hepatocyte nuclear factor 3 forkhead homolog 4|Forkhead-related protein FKHL13
- Storage Buffer:10 mM PBS, pH 7.4 with 10 mg/ml BSA and 0.1% sodium azide
- Storage Temperature:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at –20 °C, avoid freeze/thaw cycles.
- Concentration:0.50 mg/ml
- Purification:Antigen affinity purification
- Cat. No.:89282-366
- Supplier no.:GTX37424
Rabbit polyclonal antibody to FOXJ1
P-ELISA:1:500-1000, WB:1:100-500, IHC-P:1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: FOXJ1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Dog, Rat, Bovine